4-(1H-Pyrazol-1-ylmethyl)benzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 1-[4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-benzyl]-1H-pyrazole, 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)-1H-pyrazole

A364781-1G A364781-250MG A364781-5G
77240-164EA 45 USD
77240-164 77240-166 77240-168
4-(1H-Pyrazol-1-ylmethyl)benzeneboronic acid pinacol ester 98%
4-(1H-Pyrazol-1-ylmethyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₁BN₂O₂
MDL Number: MFCD18827483
CAS Number: 1315281-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR