Zoledronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (1-Hydroxy-2-(1H-imidazol-1-yl)ethane-1,1-diyl)diphosphonic acid, ZOL-446, CGP 4244, CGP 42446A, Zoledronate

A364741-100MG A364741-1G A364741-250MG A364741-50MG A364741-5G
77290-810EA 6.18 USD
77290-810 77290-812 77290-814 77290-816 77290-818
Zoledronic acid 98%
Zoledronic acid

Precaución: for Research Use Only

Formula: C₅H₁₀N₂O₇P₂
MW: 272.09 g/mol
MDL Number: MFCD00867791
CAS Number: 118072-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR