2-Amino-4'-fluorobenzophenone 95%

Supplier: Ambeed
Warning

Synonyms: (2-Aminophenyl)(4-fluorophenyl)methanone

A364661-100G A364661-10G A364661-25G A364661-5G
77251-692EA 53.56 USD
77251-692 77251-694 77251-696 77251-698
2-Amino-4'-fluorobenzophenone 95%
2-Amino-4'-fluorobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₁₀FNO
MW: 215.2230 g/mol
MDL Number: MFCD06658166
CAS Number: 3800-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR