(1-Naphthoxy)acetic acid 98%

Supplier: Ambeed
Warning

A364430-100G A364430-1G A364430-25G A364430-5G
77240-156EA 430 USD
77240-156 77240-158 77240-160 77240-162
(1-Naphthoxy)acetic acid 98%
(1-Naphthoxy)acetic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₃
MDL Number: MFCD00003927
CAS Number: 2976-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR