9-(4-Bromophenyl)-3,6-di-tert-butylcarbazole 97%

Supplier: Ambeed
Warning

A363941-1G A363941-250MG A363941-5G
77240-148EA 9.27 USD
77240-148 77240-150 77240-152 77240-154
9-(4-Bromophenyl)-3,6-di-tert-butylcarbazole 97%
9-(4-Bromophenyl)-3,6-di-tert-butylcarbazole

Precaución: for Research Use Only

Formula: C₂₆H₂₈BrN
MDL Number: MFCD11617971
CAS Number: 601454-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR