5-Amino-3-(4-chlorophenyl)isoxazole 98%

Supplier: Ambeed
Danger

Synonyms: 3-(4-Chlorophenyl)isoxazol-5-amine

A363843-100MG A363843-1G A363843-250MG A363843-5G
77278-366EA 36.05 USD
77278-366 77278-368 77278-370 77278-372
5-Amino-3-(4-chlorophenyl)isoxazole 98%
5-Amino-3-(4-chlorophenyl)isoxazole

Precaución: for Research Use Only

Formula: C₉H₇ClN₂O
MW: 194.6177 g/mol
MDL Number: MFCD01569450
CAS Number: 33866-48-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR