(2R,5S,13aR)-8-Hydroxy-7,9-dioxo-N-(2,4,6-trifluorobenzyl)-2,3,4,5,7,9,13,13a-octahydro-2,5-methanopyrido[1',2':4,5]pyrazino[2,1-b][1,3]oxazepine-10-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: Bictegravir

A363683-1G A363683-1MG A363683-5G A363683-5MG A363683-10MG A363683-100MG A363683-250MG
77264-022EA 114.33 USD
77264-022 77264-024 77264-026 77264-028 77568-744 77616-592 77616-594
(2R,5S,13aR)-8-Hydroxy-7,9-dioxo-N-(2,4,6-trifluorobenzyl)-2,3,4,5,7,9,13,13a-octahydro-2,5-methanopyrido[1',2':4,5]pyrazino[2,1-b][1,3]oxazepine-10-carboxamide 98%
(2R,5S,13aR)-8-Hydroxy-7,9-dioxo-N-(2,4,6-trifluorobenzyl)-2,3,4,5,7,9,13,13a-octahydro-2,5-methanopyrido[1',2':4,5]pyrazino[2,1-b][1,3]oxazepine-10-carboxamide

Precaución: For research use only.

Formula: C₂₁H₁₈F₃N₃O₅
MW: 449.38 g/mol
MDL Number: MFCD31536971
CAS Number: 1611493-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR