9-Butyl-6-chloro-9H-purine 97%

Supplier: Ambeed
Warning

A363461-100MG A363461-1G A363461-250MG
77251-676EA 33.99 USD
77251-676 77251-678 77251-680
9-Butyl-6-chloro-9H-purine 97%
9-Butyl-6-chloro-9H-purine

Precaución: for Research Use Only

Formula: C₉H₁₁ClN₄
MW: 210.66 g/mol
MDL Number: MFCD22207693
CAS Number: 5444-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR