2-Cyano-3-phenylpropionic acid 98%

Supplier: Ambeed
Warning

A363424-1G A363424-250MG
77264-016EA 105 USD
77264-016 77264-018
2-Cyano-3-phenylpropionic acid 98%
2-Cyano-3-phenylpropionic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MDL Number: MFCD00205073
CAS Number: 5331-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR