4-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester, 4-(Morpholine-4-carbonyl)phenylboronic Acid Pinacol Ester

A363302-100MG A363302-1G A363302-250MG
77290-790EA 16.48 USD
77290-790 77290-792 77290-794
4-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester 98%
4-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₄BNO₄
MW: 317.19 g/mol
MDL Number: MFCD05863913
CAS Number: 656239-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR