9,9-Dihexylfluorene 98%

Supplier: Ambeed
Warning

Synonyms: 9,9-Dihexyl-9H-fluorene

A363076-1G A363076-250MG A363076-5G
77251-668EA 53.56 USD
77251-668 77251-670 77251-672
9,9-Dihexylfluorene 98%
9,9-Dihexylfluorene

Precaución: for Research Use Only

Formula: C₂₅H₃₄
MW: 334.55 g/mol
MDL Number: MFCD14584648
CAS Number: 123863-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR