GSK-2982772 ≥99%

Supplier: Ambeed
Warning

Synonyms: 5-benzyl-N-[(3S)-5-methyl-4-oxo-2,3-dihydro-1,5-benzoxazepin-3-yl]-1H-1,2,4-triazole-3-carboxamide, GSK2982772, (S)-3-Benzyl-N-(5-methyl-4-oxo-2,3,4,5-tetrahydrobenzo[b][1,4]oxazepin-3-yl)-1H-1,2,4-triazole-5-carboxamide

A363017-1MG A363017-25MG A363017-5MG A363017-50MG A363017-10MG
77251-662EA 28 USD
77251-662 77251-664 77251-666 77746-722 77746-721
GSK-2982772 ≥99%
GSK-2982772

Precaución: for Research Use Only

Formula: C₂₀H₁₉N₅O₃
MDL Number: MFCD30828708
CAS Number: 1622848-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR