2-Cyano-6-methoxybenzothiazole 96%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxy-2-benzothiazolecarbonitrile, 6-Methoxybenzo[d]thiazole-2-carbonitrile

A362687-100MG A362687-1G A362687-250MG A362687-5G
77264-008EA 5.15 USD
77264-008 77264-010 77264-012 77264-014
2-Cyano-6-methoxybenzothiazole 96%
2-Cyano-6-methoxybenzothiazole

Precaución: for Research Use Only

Formula: C₉H₆N₂OS
MW: 190.22 g/mol
MDL Number: MFCD00010537
CAS Number: 943-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR