Boc-L-Homoallylglycine Methyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Boc-HoAllyl-Gly-OMe

A362665-250MG A362665-5G A362665-1G
77240-136EA 85 USD
77240-136 77551-044 77568-740
Boc-L-Homoallylglycine Methyl ester 97%
Boc-L-Homoallylglycine Methyl ester

Precaución: For research use only.

Formula: C₁₂H₂₁NO₄
MW: 243.29 g/mol
MDL Number: MFCD01632226
CAS Number: 92136-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR