Ethyl-4-(4-bromophenyl)-4-oxobutyrate 96%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-(4-bromophenyl)-4-oxobutanoate

A362471-100G A362471-10G A362471-1G A362471-250MG A362471-25G A362471-5G
77290-778EA 425.39 USD
77290-778 77290-780 77290-782 77290-784 77290-786 77290-788
Ethyl-4-(4-bromophenyl)-4-oxobutyrate 96%
Ethyl-4-(4-bromophenyl)-4-oxobutyrate

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrO₃
MW: 285.14 g/mol
MDL Number: MFCD00672068
CAS Number: 30913-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR