4,5-Difluoro-2-nitrophenol 98%

Supplier: Ambeed
Danger

A362468-1G A362468-5G A362468-25G
77278-352EA 19 USD
77278-352 77278-354 77616-590
4,5-Difluoro-2-nitrophenol 98%
4,5-Difluoro-2-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃F₂NO₃
MDL Number: MFCD03094187
CAS Number: 55346-97-9
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR