2,3-Dihydroxy-4-methoxybenzaldehyde 95%

Supplier: Ambeed
Warning

A362192-100MG A362192-250MG
77251-658EA 54 USD
77251-658 77251-660
2,3-Dihydroxy-4-methoxybenzaldehyde 95%
2,3-Dihydroxy-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₄
MDL Number: MFCD00016589
CAS Number: 4055-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR