3-Chloro-5-(trifluoromethyl)picolinic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Chloro-5-(trifluoromethyl)-2-pyridinecarboxylic acid, 3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid, 3-Chloro-5-(trifluoromethyl)picolinic acid
A361848-100G
A361848-10G
A361848-1G
A361848-25G
A361848-5G
77278-320EA
298.7
USD
77278-320
77278-322
77278-324
77278-326
77278-328
3-Chloro-5-(trifluoromethyl)picolinic acid 98%
3-Chloro-5-(trifluoromethyl)picolinic acid
Precaución: for Research Use Only
Formula:
C₇H₃ClF₃NO₂ MW: 225.60 g/mol |
MDL Number:
MFCD00277482 CAS Number: 80194-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...