3-Amino-1H-1,2,4-triazole-5-carboxylic acid hydrate 98%

Supplier: Ambeed
Warning

Synonyms: 1H-1,2,4-Triazole-3-carboxylicacid, 5-amino-, hydrate (1:1), 3-Amino-1,2,4-triazole-5-carboxylic acid hydrate, 5-Amino-2H-1, 2, 4-triazole-3-carboxylic acid hydrate

A361230-25G A361230-5G
77278-316EA 58.71 USD
77278-316 77278-318
3-Amino-1H-1,2,4-triazole-5-carboxylic acid hydrate 98%
3-Amino-1H-1,2,4-triazole-5-carboxylic acid hydrate

Precaución: for Research Use Only

Formula: C₃H₄N₄O₂·H₂O
MW: 146.10 g/mol
MDL Number: MFCD00149595
CAS Number: 304655-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR