N-(tert-Butyl)-1-(1-(5-(5-(trifluoromethyl)-1,3,4-oxadiazol-2-yl)pyridin-2-yl)piperidin-4-yl)-1H-imidazole-5-carboxamide 98%
Supplier: Ambeed
|
Warning
|
A361136-10MG
A361136-1MG
A361136-25MG
A361136-50MG
A361136-5MG
77263-986EA
61.8
USD
77263-986
77263-988
77263-990
77263-992
77263-994
N-(tert-Butyl)-1-(1-(5-(5-(trifluoromethyl)-1,3,4-oxadiazol-2-yl)pyridin-2-yl)piperidin-4-yl)-1H-imidazole-5-carboxamide 98%
N-(tert-Butyl)-1-(1-(5-(5-(trifluoromethyl)-1,3,4-oxadiazol-2-yl)pyridin-2-yl)piperidin-4-yl)-1H-imidazole-5-carboxamide
Precaución: for Research Use Only
Formula:
C₂₁H₂₄F₃N₇O₂ MW: 463.45 g/mol |
MDL Number:
MFCD30489744 CAS Number: 2012607-27-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...