1-Iodo-2-(trifluoromethoxy)benzene 98%

Supplier: Ambeed
Warning

A361092-10G A361092-1G A361092-25G A361092-5G A361092-100G
77251-650EA 8 USD
77251-650 77251-652 77251-654 77251-656 77746-718
1-Iodo-2-(trifluoromethoxy)benzene 98%
1-Iodo-2-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₄F₃IO
MDL Number: MFCD00042410
CAS Number: 175278-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR