tert-Butyl (3-amino-2,2-dimethylpropyl)carbamate 98%

Supplier: Ambeed
Warning

A360949-250MG A360949-5G A360949-1G
77240-134EA 13 USD
77240-134 77594-072 77746-714
tert-Butyl (3-amino-2,2-dimethylpropyl)carbamate 98%
tert-Butyl (3-amino-2,2-dimethylpropyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₂₂N₂O₂
MDL Number: MFCD11227269
CAS Number: 292606-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR