2-(3,5-Dichlorophenyl)acetonitrile 98%

Supplier: Ambeed
Warning

A360873-10G A360873-1G A360873-25G A360873-5G
77240-116EA 37 USD
77240-116 77240-118 77240-120 77240-122
2-(3,5-Dichlorophenyl)acetonitrile 98%
2-(3,5-Dichlorophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₅Cl₂N
MDL Number: MFCD09923674
CAS Number: 52516-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR