4,5-Difluoro-2-methylphenol 95%

Supplier: Ambeed
Warning

A360766-100MG A360766-1G
77290-768EA 26.78 USD
77290-768 77748-719
4,5-Difluoro-2-methylphenol 95%
4,5-Difluoro-2-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆F₂O
MW: 144.12 g/mol
MDL Number: MFCD04972820
CAS Number: 704884-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR