2-Di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl 98%

Supplier: Ambeed
Warning

Synonyms: t-BuXPhos, Di-tert-butyl(2',4',6'-triisopropyl-[1,1'-biphenyl]-2-yl)phosphine, 2-Di-tert-butylphosphino-2',4',6'-triisopropylbiphenyl, tBuXPhos

A360721-100G A360721-1G A360721-250MG A360721-25G A360721-10G A360721-5G
77263-978EA 236 USD
77263-978 77263-980 77263-982 77263-984 77551-032 77551-034
2-Di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl 98%
2-Di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₂₉H₄₅P
MW: 424.65 g/mol
MDL Number: MFCD06411306
CAS Number: 564483-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR