(±)-3-Hydroxybutyric acid 80%

Supplier: Ambeed
Warning

A360305-100G A360305-1G A360305-25G A360305-5G
77278-284EA 540 USD
77278-284 77278-286 77278-288 77278-290
(±)-3-Hydroxybutyric acid 80%
(±)-3-Hydroxybutyric acid

Precaución: for Research Use Only

Formula: C₄H₈O₃
MDL Number: MFCD00004546
CAS Number: 300-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR