10-Bromonaphtho[2,1-b]benzofuran 98%

Supplier: Ambeed
Warning

A360233-1G A360233-250MG A360233-5G
77263-972EA 45 USD
77263-972 77263-974 77263-976
10-Bromonaphtho[2,1-b]benzofuran 98%
10-Bromonaphtho[2,1-b]benzofuran

Precaución: for Research Use Only

Formula: C₁₆H₉BrO
MDL Number: MFCD30470213
CAS Number: 1256544-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR