4'-Fluoro-3'-methoxyacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Fluoro-3-methoxyphenyl)ethanone

A359695-100MG A359695-1G A359695-250MG A359695-25G A359695-5G
77278-266EA 16 USD
77278-266 77278-268 77278-270 77278-272 77278-274
4'-Fluoro-3'-methoxyacetophenone 98%
4'-Fluoro-3'-methoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MDL Number: MFCD00272135
CAS Number: 64287-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR