3-Amino-4-hydroxybenzonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-4-hydroxybenzonitrile, 2-Amino-4-cyanophenol, 2-Amino-4-cyano-phenol

A359128-10G A359128-1G A359128-250MG A359128-25G A359128-5G
77251-640EA 111 USD
77251-640 77251-642 77251-644 77251-646 77572-056
3-Amino-4-hydroxybenzonitrile 97%
3-Amino-4-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MDL Number: MFCD00234273
CAS Number: 14543-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR