(±)-1-Phenyl-1-propanol 98%

Supplier: Ambeed
Warning

A358826-100G A358826-25G A358826-500G A358826-5G
77240-100EA 38 USD
77240-100 77240-102 77240-104 77240-106
(±)-1-Phenyl-1-propanol 98%
(±)-1-Phenyl-1-propanol

Precaución: for Research Use Only

Formula: C₆H₅CH(OH)CH₂CH₃
MDL Number: MFCD00004564
CAS Number: 93-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR