NCB-0846 99+%

Supplier: Ambeed
Warning

Synonyms: cis-4-((2-((1H-Benzo[d]imidazol-6-yl)amino)quinazolin-8-yl)oxy)cyclohexanol

A358703-10MG A358703-1MG A358703-5MG
77263-962EA 112 USD
77263-962 77263-964 77263-966
NCB-0846 99+%
NCB-0846

Precaución: for Research Use Only

Formula: C₂₁H₂₁N₅O₂
MW: 375.42 g/mol
MDL Number: MFCD30749187
CAS Number: 1792999-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR