4-Ethylaniline 98%

Supplier: Ambeed
Danger

A358292-100G A358292-10G A358292-25G A358292-500G A358292-5G
77290-736EA 33.99 USD
77290-736 77290-738 77290-740 77290-742 77290-744
4-Ethylaniline 98%
4-Ethylaniline

Precaución: for Research Use Only

Formula: C₈H₁₁N
MW: 121.18 g/mol
MDL Number: MFCD00007921
CAS Number: 589-16-2
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR