4-(1-Pyrrolidinyl)benzenecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(1-Pyrrolidinyl)benzoic acid, 4-(Pyrrolidin-1-yl)benzoic acid

A358275-1G A358275-250MG
77290-732EA 36.05 USD
77290-732 77290-734
4-(1-Pyrrolidinyl)benzenecarboxylic acid 98%
4-(1-Pyrrolidinyl)benzenecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₂
MW: 191.23 g/mol
MDL Number: MFCD01631241
CAS Number: 22090-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR