1-(2-Propynyl)piperazine 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Prop-2-yn-1-yl)piperazine, 1-Propargylpiperazine

A358203-100MG A358203-1G A358203-250MG
77240-094EA 17 USD
77240-094 77240-096 77240-098
1-(2-Propynyl)piperazine 98%
1-(2-Propynyl)piperazine

Precaución: for Research Use Only

Formula: C₇H₁₂N₂
MW: 124.19 g/mol
MDL Number: MFCD08445213
CAS Number: 52070-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR