4'-Benzyloxy-2'-hydroxyacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-(Benzyloxy)-2-hydroxyphenyl)ethanone

A357871-100MG A357871-10G A357871-1G A357871-250MG A357871-25G A357871-5G
77290-718EA 7.21 USD
77290-718 77290-720 77290-722 77290-724 77290-726 77290-728
4'-Benzyloxy-2'-hydroxyacetophenone 98%
4'-Benzyloxy-2'-hydroxyacetophenone

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.2699 g/mol
MDL Number: MFCD00054612
CAS Number: 29682-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR