Dicarbonylcyclopentadienylruthenium dimer 98%

Supplier: Ambeed
Warning

A357400-100MG A357400-250MG A357400-1G
77251-624EA 79 USD
77251-624 77251-626 77616-568
Dicarbonylcyclopentadienylruthenium dimer 98%
Dicarbonylcyclopentadienylruthenium dimer

Precaución: for Research Use Only

Formula: C₁₄H₁₀O₄Ru₂
MW: 444.3700 g/mol
MDL Number: MFCD00070447
CAS Number: 12132-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR