Bis(2,5-dioxopyrrolidin-1-yl) adipate 97%

Supplier: Ambeed
Warning

Synonyms: Di(N-succinimidyl)adipate

A357319-1G A357319-250MG A357319-5G
77263-952EA 46 USD
77263-952 77263-954 77616-566
Bis(2,5-dioxopyrrolidin-1-yl) adipate 97%
Bis(2,5-dioxopyrrolidin-1-yl) adipate

Precaución: for Research Use Only

Formula: C₁₄H₁₆N₂O₈
MDL Number: MFCD01318516
CAS Number: 59156-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR