BV6 98%

Supplier: Ambeed
Warning

Synonyms: (2S,2'S)-N,N'-((2S,2'S)-(Hexane-1,6-diylbis(azanediyl))bis(1-oxo-3,3-diphenylpropane-1,2-diyl))bis(1-((S)-2-cyclohexyl-2-((S)-2-(methylamino)propanamido)acetyl)pyrrolidine-2-carboxamide), (S,S,2S,2'S)-N,N'-((2S,2'S)-(hexane-1,6-diylbis(azanediyl))bis(3-oxo-1,1-diphenylpropane-3,2-diyl))bis(1-((S)-2-cyclohexyl-2-((S)-2-(methylamino)

A357114-1MG A357114-2MG A357114-50MG A357114-5MG
77240-084EA 25 USD
77240-084 77240-086 77240-088 77240-090
BV6 98%
BV6

Precaución: for Research Use Only

Formula: C₇₀H₉₆N₁₀O₈
MDL Number: MFCD28168021
CAS Number: 1001600-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR