1-Methyl-4-prop-2-ynyl-piperazine 98%

Supplier: Ambeed
Danger

A357043-250MG A357043-1G A357043-100MG
77240-080EA 74.16 USD
77240-080 77616-564 77746-703
1-Methyl-4-prop-2-ynyl-piperazine 98%
1-Methyl-4-prop-2-ynyl-piperazine

Precaución: for Research Use Only

Formula: C₈H₁₄N₂
MW: 138.21 g/mol
MDL Number: MFCD09910338
CAS Number: 45813-02-3
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR