1-Chloro-3-(3,4-dimethoxyphenyl)-3-oxopropane 97%

Supplier: Ambeed
Danger

A356823-100MG A356823-1G A356823-250MG
77278-238EA 68 USD
77278-238 77278-240 77278-242
1-Chloro-3-(3,4-dimethoxyphenyl)-3-oxopropane 97%
1-Chloro-3-(3,4-dimethoxyphenyl)-3-oxopropane

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₃
MDL Number: MFCD00510412
CAS Number: 4693-38-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR