4-((Ethoxycarbonyl)amino)benzoic acid 97%

Supplier: Ambeed
Warning

A356635-100MG A356635-1G A356635-250MG
77290-708EA 28.84 USD
77290-708 77290-710 77290-712
4-((Ethoxycarbonyl)amino)benzoic acid 97%
4-((Ethoxycarbonyl)amino)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₄
MW: 209.19 g/mol
MDL Number: MFCD00610504
CAS Number: 5180-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR