3,3',5,5'-Tetrakis(methoxymethyl)-[1,1'-biphenyl]-4,4'-diol 97%

Supplier: Ambeed
Warning

A356457-1G A356457-250MG A356457-25G A356457-5G
77240-072EA 16.48 USD
77240-072 77240-074 77240-076 77240-078
3,3',5,5'-Tetrakis(methoxymethyl)-[1,1'-biphenyl]-4,4'-diol 97%
3,3',5,5'-Tetrakis(methoxymethyl)-[1,1'-biphenyl]-4,4'-diol

Precaución: for Research Use Only

Formula: C₂₀H₂₆O₆
MW: 362.41 g/mol
MDL Number: MFCD30187312
CAS Number: 455943-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR