ω-Bromo-4-methoxyacetophenone 98%

Supplier: Ambeed
Danger

Synonyms: 4-Methoxyphenacyl bromide, Bromomethyl p-methoxyphenyl ketone, α-Bromo-4-methoxyacetophenone, 2-Bromo-1-(4-methoxyphenyl)ethan-1-one, 2-Bromo-1-(4-methoxyphenyl)ethanone, 2-Bromo-4'-methoxyacetophenone

A356070-100G A356070-10G A356070-25G A356070-25MG A356070-5G A356070-5MG
77263-924EA 70.04 USD
77263-924 77263-926 77263-928 77263-930 77263-932 77263-934
ω-Bromo-4-methoxyacetophenone 98%
ω-Bromo-4-methoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
MDL Number: MFCD00000201
CAS Number: 2632-13-5
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR