4-Hydroxy-3-methoxypropiophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Hydroxy-3-methoxyphenyl)propan-1-one

A356046-100MG A356046-10G A356046-1G A356046-250MG A356046-5G
77290-692EA 31.93 USD
77290-692 77290-694 77290-696 77290-698 77290-700
4-Hydroxy-3-methoxypropiophenone 98%
4-Hydroxy-3-methoxypropiophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
MDL Number: MFCD00482070
CAS Number: 1835-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR