5-Amino-2-(4-aminophenyl)benzimidazole 98%

Supplier: Ambeed
Danger

Synonyms: 2-(4-Aminophenyl)-1H-benzimidazol-5-amine

A355877-1G A355877-25000G A355877-25G
77240-068EA 7.21 USD
77240-068 77442-226 77551-010
5-Amino-2-(4-aminophenyl)benzimidazole 98%
5-Amino-2-(4-aminophenyl)benzimidazole

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₄
MW: 224.27 g/mol
MDL Number: MFCD00451475
CAS Number: 7621-86-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR