1,1'-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]ferrocene ≥97%, ee 99%

Supplier: Ambeed
Warning

Synonyms: 1,1-Bis((2R,5R)-2,5-di-i-propylphospholano)ferrocene, (R,R)-Isopropyl-Ferrocelane

A355614-100MG A355614-10G A355614-1G A355614-5G A355614-250MG
77251-594EA 51.5 USD
77251-594 77251-596 77251-598 77251-600 77616-560
1,1'-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]ferrocene ≥97%, ee 99%
1,1'-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]ferrocene

Precaución: for Research Use Only

Formula: C₃₀H₄₀FeP₂
MW: 518.43 g/mol
MDL Number: MFCD09750465
CAS Number: 849950-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR