3,6-Dibromo-2,7-diiodophenanthrene-9,10-dione 95%

Supplier: Ambeed
Warning

A355305-100MG A355305-1G A355305-250MG A355305-5G
77240-054EA 25 USD
77240-054 77240-056 77240-058 77616-558
3,6-Dibromo-2,7-diiodophenanthrene-9,10-dione 95%
3,6-Dibromo-2,7-diiodophenanthrene-9,10-dione

Precaución: for Research Use Only

Formula: C₁₄H₄Br₂I₂O₂
MDL Number: MFCD30342486
CAS Number: 887127-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR