(4,4',5,5'-Tetrachloro-1'H-[1,3'-bipyrrole]-2,2'-diyl)bis((2-hydroxyphenyl)methanone) 97%
Supplier: Ambeed
|
Danger
|
A354912-10MG
A354912-1MG
A354912-2MG
A354912-5MG
77240-038EA
62
USD
77240-038
77240-040
77240-042
77240-044
(4,4',5,5'-Tetrachloro-1'H-[1,3'-bipyrrole]-2,2'-diyl)bis((2-hydroxyphenyl)methanone) 97%
(4,4',5,5'-Tetrachloro-1'H-[1,3'-bipyrrole]-2,2'-diyl)bis((2-hydroxyphenyl)methanone)
Precaución: for Research Use Only
Formula:
C₂₂H₁₂Cl₄N₂O₄ |
CAS Number:
1227962-62-0 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...