2-Ethyl-2-hexenal (cis- and trans- mixture) 94%

Supplier: Ambeed
Danger

A354763-25G A354763-5G
77263-916EA 117 USD
77263-916 77263-918
2-Ethyl-2-hexenal (cis- and trans- mixture) 94%
2-Ethyl-2-hexenal (cis- and trans- mixture)

Precaución: for Research Use Only

Formula: C₈H₁₄O
MDL Number: MFCD00006983
CAS Number: 645-62-5
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR