3-Fluoro-(5-methoxycarbonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-5-methoxycarbonylphenylboronic Acid, 3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid

A354371-1G A354371-250MG A354371-5G
77240-030EA 14.42 USD
77240-030 77240-032 77240-034 77240-036
3-Fluoro-(5-methoxycarbonyl)phenylboronic acid 98%
3-Fluoro-(5-methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BFO₄
MW: 197.96 g/mol
MDL Number: MFCD07363749
CAS Number: 871329-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR